Få mere at vide
(2S,4S)-4-Methylglutamic Acid, TRC
Artikelnummer. 30482737
Skift visning
Mængde:
5 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30482737 10 mg
30465731 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30482737 Leverandør Toronto Research Chemicals Leverandørnr. TRCM31158010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 6141-27-1
Molekylær formel C6 H11 N O4
Molekylvægt (g/mol) 161.16
IUPAC navn (2S,4S)-2-amino-4-methylpentanedioic acid
SMIL C[C@@H](C[C@H](N)C(=O)O)C(=O)O
Tekniske data
Kemisk navn eller materiale (2S,4S)-4-Methylglutamic Acid
Mængde 10 mg
InChI formel InChI=1S/C6H11NO4/c1-3(5(8)9)2-4(7)6(10)11/h3-4H,2,7H2,1H3,(H,8,9)(H,10,11)/t3-,4-/m0/s1
Formel vægt 161.0688