Få mere at vide
(2S,4R)-4-hydroxypyrrolidin-1,2-dicarboxylsyre 1-tert-butylester 2-methylester, TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | (2S,4R)-4-Hydroxypyrrolidine-1,2-dicarboxylic Acid 1-tert-butyl ester 2-methyl ester |
| CAS | 74844-91-0 |
| Molekylær formel | C11 H19 N O5 |
| Mængde | 1 g |
| Synonym | 1,2-Pyrrolidinedicarboxylic acid, 4-hydroxy-, 1-(1,1-dimethylethyl) 2-methyl ester, (2S,4R)-, 1,2-Pyrrolidinedicarboxylic acid, 4-hydroxy-, 1-(1,1-dimethylethyl) 2-methyl ester, (2S-trans)-, (2S,4R)-1-tert-Butoxycarbonyl-2-(methoxycarbonyl)-4-hydroxypyrrolidine, (2S,4R)-1-tert-Butoxycarbonyl-4-hydroxypyrrolidine-2-carboxylic acid methyl ester, (2S,4R)-4-Hydroxypyrrolidine-1,2-dicarboxylic acid 1-tert-butyl ester 2-methyl ester, (2S,4R)-Methyl 1-(tert-butyloxycarbonyl)-4-hydroxypyrrolidine-2-carboxylate, 1-tert-Butyl 2-methyl (2S,4R)-4-hydroxy-1,2-pyrrolidinedicarboxylate, 1-tert-Butyl 2-methyl 4(R)-hydroxypyrrolidine-1,2(S)-dicarboxylate, Methyl (2S,4R)-1-(tert-butoxycarbonyl)-4-hydroxypyrrolidine-2-carboxylate, Methyl (2S,4R)-N-tert-Butoxycarbonyl-4-hydroxy-2-pyrrolidinecarboxylate, Methyl (2S,4R)-N-tert-butoxycarbonyl-4-hydroxy-2-pyrrolidinecarboxylate, Methyl (4R)-N-Boc-4-hydroxy-L-prolinate, Methyl N-tert-butoxycarbonyl-L-hydroxyprolinate, Methyl-(N-Boc-4R-hydroxypyrrolidine)-2-carboxylate, N-(tert-Butoxycarbonyl)-(2S,4R)-4-hydroxyproline methyl ester, N-(tert-Butoxycarbonyl)-(2S,4R)-4-hydroxyproline methyl ester, N-Boc-4-hydroxy-L-proline methyl ester, N-Boc-trans-4-hydroxyproline methyl ester, N-tert-Butoxycarbonyl-L-hydroxyproline methyl ester, tert-Butyl (2S,4R)-2-(methoxycarbonyl)-4-hydroxypyrrolidine-1-carboxylate, tert-Butyl methyl (2S,4R)-4-hydroxypyrrolidine-1,2-dicarboxylate |
| InChI formel | InChI=1S/C11H19NO5/c1-11(2,3)17-10(15)12-6-7(13)5-8(12)9(14)16-4/h7-8,13H,5-6H2,1-4H3/t7-,8+/m1/s1 |
| SMIL | COC(=O)[C@@H]1C[C@@H](O)CN1C(=O)OC(C)(C)C |
| IUPAC navn | 1-O-tert-butyl 2-O-methyl (2S,4R)-4-hydroxypyrrolidin-1,2-dicarboxylat |
| Molekylvægt (g/mol) | 245.27 |
| Formel vægt | 245.1263 |