Få mere at vide
(2S,3S)-3-Hydroxy-2-(4-methoxyphenyl)-2,3-dihydro-1,5-benzothiazepin-4(5H)-one, Mikromol™
Artikelnummer. 30051168
Skift visning
Mængde:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30051168 100 mg
30049248 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30051168 Leverandør Mikromol Leverandørnr. MM0068.04
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
(2S,3S)-3-Hydroxy-2-(4-methoxyphenyl)-2,3-dihydro-1,5-benzothiazepin-4(5H)-one
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale (2S,3S)-3-Hydroxy-2-(4-methoxyphenyl)-2,3-dihydro-1,5-benzothiazepin-4(5H)-one
CAS 42399-49-5
Analyt- eller komponentnavne (2S,3S)-3-Hydroxy-2-(4-methoxyphenyl)-2,3-dihydro-1,5-benzothiazepin-4(5H)-one
Forsendelsestilstand Room Temperature
Molekylær formel C16 H15 N O3 S
Certificeringer/compliance ISO 17025
Synonym Diltiazem Hydrochloride Imp. E (EP), Diltiazem Imp. E (EP), (2S,3S)-3-Hydroxy-2-(4-methoxyphenyl)-2,3-dihydro-1,5-benzothiazepin-4(5H)-one, Diltiazem Hydrochloride Impurity E, Diltiazem Impurity E
InChI formel InChI=1S/C16H15NO3S/c1-20-11-8-6-10(7-9-11)15-14(18)16(19)17-12-4-2-3-5-13(12)21-15/h2-9,14-15,18H,1H3,(H,17,19)/t14-,15+/m1/s1
SMIL COc1ccc(cc1)[C@@H]2Sc3ccccc3NC(=O)[C@@H]2O
IUPAC navn (2S,3S)-3-hydroxy-2-(4-methoxyphenyl)-3,5-dihydro-2H-1,5-benzothiazepin-4-one
Molekylvægt (g/mol) 301.36
Mængde 100 mg
Formel vægt 301.0773 g/mol