missing translation for 'onlineSavingsMsg'
Få mere at vide

(2S,3S)-3-(2-Aminophenyl)sulfanyl-2-hydroxy-3-(4-methoxyphenyl)propanoic Acid, Mikromol™

Product Code. 30052078
Change view
Click to view available options
Mængde:
25 mg
100 mg
4 x 25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
30052078 25 mg
30052052 100 mg
30460793 4 x 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30052078 Supplier Mikromol Supplier No. MM0068.130025

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

(2S,3S)-3-(2-Aminophenyl)sulfanyl-2-hydroxy-3-(4-methoxyphenyl)propanoic Acid
TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale (2S,3S)-3-(2-Aminophenyl)sulfanyl-2-hydroxy-3-(4-methoxyphenyl)propanoic Acid
CAS 42399-48-4
Analyt- eller komponentnavne (2S,3S)-3-(2-Aminophenyl)sulfanyl-2-hydroxy-3-(4-methoxyphenyl)propanoic Acid
Forsendelsestilstand Room Temperature
Molekylær formel C16 H17 N O4 S
Certificeringer/compliance ISO 17025
Synonym (2S,3S)-3-(2-Aminophenyl)sulfanyl-2-hydroxy-3-(4-methoxyphenyl)propanoic Acid, (alphaS,betaS)-beta-[(2-Aminophenyl)thio]-alpha-hydroxy-4-methoxybenzenepropanoic acid, Benzenepropanoic acid, beta-[(2-aminophenyl)thio]-alpha-hydroxy-4-methoxy-, [S-(R*,R*)]-, d-threo-2-Hydroxy-3-(2-aminophenylthio)-3-(4-methoxyphenyl)propionic acid
InChI formel InChI=1S/C16H17NO4S/c1-21-11-8-6-10(7-9-11)15(14(18)16(19)20)22-13-5-3-2-4-12(13)17/h2-9,14-15,18H,17H2,1H3,(H,19,20)/t14-,15+/m1/s1
SMIL COc1ccc(cc1)[C@H](Sc2ccccc2N)[C@@H](O)C(=O)O
IUPAC navn (2S,3S)-3-(2-aminophenyl)sulfanyl-2-hydroxy-3-(4-methoxyphenyl)propanoic acid
Molekylvægt (g/mol) 319.38
Mængde 25 mg
Formel vægt 319.0878 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.