missing translation for 'onlineSavingsMsg'
Få mere at vide

(2S,3S)-2-Amino-3-methyl-N-[2-(4-morpholinyl)ethyl]pentanamide Hydrochloride, TRC

Artikelnummer. 30323369
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
2.5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30323369 2.5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30323369 Leverandør Toronto Research Chemicals Leverandørnr. TRCA6125002.5G

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Tekniske data

Kemisk navn eller materiale (2S,3S)-2-Amino-3-methyl-N-[2-(4-morpholinyl)ethyl]pentanamide Hydrochloride
CAS 1243259-19-9
Molekylær formel C12H25N3O2 . 2HCl
Mængde 2.5 g
Synonym [S-(R*,R*)]-2-Amino-3-methyl-N-[2-(4-morpholinyl)ethyl]pentanamide Dihydrochloride, LM11A 31 Dihydrochloride
InChI formel InChI=1S/C12H25N3O2/c1-3-10(2)11(13)12(16)14-4-5-15-6-8-17-9-7-15/h10-11H,3-9,13H2,1-2H3,(H,14,16)/t10-,11-/m0/s1
SMIL Cl.Cl.CC[C@H](C)[C@H](N)C(=O)NCCN1CCOCC1
IUPAC navn (2S,3S)-2-amino-3-methyl-N-(2-morpholin-4-ylethyl)pentanamide
Molekylvægt (g/mol) 243.35
Formel vægt 315.148
Produkttitel
Vælg et problem

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.