Få mere at vide
(2S,3aR,7aS)-Octahydroindole-2-carboxylic Acid (~90%, contains cis), TRC
Artikelnummer. 30428006
Skift visning
Mængde:
50 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30428006 50 mg
30411301 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30428006 Leverandør Toronto Research Chemicals Leverandørnr. TRCO23599550MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 145438-94-4
Molekylær formel C9H15NO2
Molekylvægt (g/mol) 169.22
Synonym rel-(2S, 3aR,7aS)-Octahydro-1H-indole-2-carboxylic Acid, (2alpha,3aalpha,7abeta)-Octahydro-1H-indole-2-carboxylic Acid
IUPAC navn (2S,3aR,7aS)-2,3,3a,4,5,6,7,7a-octahydro-1H-indole-2-carboxylic acid
SMIL OC(=O)[C@@H]1C[C@H]2CCCC[C@@H]2N1
Tekniske data
Kemisk navn eller materiale (2S,3aR,7aS)-Octahydroindole-2-carboxylic Acid
Mængde 50 mg
InChI formel InChI=1S/C9H15NO2/c11-9(12)8-5-6-3-1-2-4-7(6)10-8/h6-8,10H,1-5H2,(H,11,12)/t6-,7+,8+/m1/s1
Formel vægt 169.1103