missing translation for 'onlineSavingsMsg'
Learn More

(2S)- 2-Methylpyrrolidine-2-carboxylic Acid Hydrochloride, TRC

Product Code. 30415394
Change view
Click to view available options
Mængde:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
30415394 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30415394 Supplier Toronto Research Chemicals Supplier No. TRCS2352651G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale (2S)- 2-Methylpyrrolidine-2-carboxylic Acid Hydrochloride
CAS 1508261-86-6
Molekylær formel C6 H11 N O2 . H Cl
Mængde 1 g
Synonym L-Proline','2-methyl-','hydrochloride (1:1) (ACI), 2-Methyl-L-proline hydrochloride
InChI formel InChI=1S/C6H11NO2.ClH/c1-6(5(8)9)3-2-4-7-6;/h7H,2-4H2,1H3,(H,8,9);1H/t6-;/m0./s1
SMIL C[C@]1(CCCN1)C(=O)O.Cl
IUPAC navn (2S)-2-methylpyrrolidine-2-carboxylic acid;hydrochloride
Molekylvægt (g/mol) 165.61798
Formel vægt 165.0557
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.