Få mere at vide
(2S)-2-Methylbutanoic Acid (1S,3S,7S,8S,8aR)-8-[2-[(2R)-3,6-Dihydro-6-oxo-2H-pyran-2-yl]ethyl]-1,2,3,7,8,8a-hexahydro-3-hydroxy-7-methyl-1-naphthalenyl Ester, TRC
Artikelnummer. 30473375
Skift visning
Mængde:
10 mg
25 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30473375 100 mg
30479084 25 mg
30467671 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30473375 Leverandør Toronto Research Chemicals Leverandørnr. TRCM303875100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1114834-63-7
Molekylær formel C23H32O5
Molekylvægt (g/mol) 388.5
Synonym (2S)-2-Methylbutanoic acid (1S,3S,7S,8S,8aR)-8-[2-[(2R)-3,6-Dihydro-6-oxo-2H-pyran-2-yl]ethyl]-1,2,3,7,8,8a-hexahydro-3-hydroxy-7-methyl-1-naphthalenyl Ester, Anhydropravastatin Lactone
IUPAC navn [(1S,3S,7S,8S,8aR)-3-hydroxy-7-methyl-8-[2-[(2R)-6-oxo-2,3-dihydropyran-2-yl]ethyl]-1,2,3,7,8,8a-hexahydronaphthalen-1-yl] (2S)-2-methylbutanoate
SMIL CC[C@H](C)C(=O)O[C@H]1C[C@H](O)C=C2C=C[C@H](C)[C@H](CC[C@@H]3CC=CC(=O)O3)[C@@H]12
Tekniske data
Kemisk navn eller materiale (2S)-2-Methylbutanoic Acid (1S,3S,7S,8S,8aR)-8-[2-[(2R)-3,6-Dihydro-6-oxo-2H-pyran-2-yl]ethyl]-1,2,3,7,8,8a-hexahydro-3-hydroxy-7-methyl-1-naphthalenyl Ester
Mængde 100 mg
InChI formel InChI=1S/C23H32O5/c1-4-14(2)23(26)28-20-13-17(24)12-16-9-8-15(3)19(22(16)20)11-10-18-6-5-7-21(25)27-18/h5,7-9,12,14-15,17-20,22,24H,4,6,10-11,13H2,1-3H3/t14-,15-,17+,18-,19-,20-,22-/m0/s1
Formel vægt 388.225