Få mere at vide
(2S)-2-Mercaptopropanoic Acid, TRC
Artikelnummer. 30478833
Skift visning
Mængde:
50 mg
100 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30478833 100 mg
30481024 250 mg
30467313 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30478833 Leverandør Toronto Research Chemicals Leverandørnr. TRCM202725100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 57965-30-7
Molekylær formel C3H6O2S
Molekylvægt (g/mol) 106.14
Synonym (-)-2-Mercaptopropionic Acid, 2(S)-Mercaptopropanoic Acid
IUPAC navn (2S)-2-sulfanylpropanoic acid
SMIL C[C@H](S)C(=O)O
Tekniske data
Kemisk navn eller materiale (2S)-2-Mercaptopropanoic Acid
Mængde 100 mg
InChI formel InChI=1S/C3H6O2S/c1-2(6)3(4)5/h2,6H,1H3,(H,4,5)/t2-/m0/s1
Formel vægt 106.0089