Få mere at vide
(2S)-2-Hydroxyglutaric Acid Octyl Ester Sodium Salt, TRC
Artikelnummer. 30003696
Skift visning
Mængde:
5 mg
10 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30003696 5 mg
30007829 50 mg
30008168 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30003696 Leverandør Toronto Research Chemicals Leverandørnr. TRCH9425965MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1391067-96-1
Molekylær formel C13H23NaO5
Molekylvægt (g/mol) 282.31
Synonym (2S)-2-Hydroxypentanedioic Acid Octyl Ester Sodium Salt, (S)-2-Hydroxyglutaric Acid Octyl Ester Sodium Salt, (S)-α-Hydroxyglutaric Acid Octyl Ester Sodium Salt, L-2-Hydroxyglutaric Acid Octyl Ester Sodium Salt, Octyl L-2-Hydroxyglutarate Sodium Salt, Octyl-L-2-HG Sodium;
IUPAC navn sodium;(4S)-4-hydroxy-5-octoxy-5-oxopentanoate
SMIL CCCCCCCCOC(=O)[C@H](CCC(=O)[O-])O.[Na+]
Tekniske data
Kemisk navn eller materiale (2S)-2-Hydroxyglutaric Acid Octyl Ester Sodium Salt
Renhedsgrad noter HPLC
Mængde 5 mg
InChI formel InChI=1S/C13H24O5.Na/c1-2-3-4-5-6-7-10-18-13(17)11(14)8-9-12(15)16;/h11,14H,2-10H2,1H3,(H,15,16);/q;+1/p-1/t11-;/m0./s1
Formel vægt 282.1443
Procent renhed >95%