Få mere at vide
(2S)-2-Ethyl Ester-isocyanato-propanoic Acid, TRC
Artikelnummer. 30180250
Skift visning
Mængde:
50 mg
100 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30180250 100 mg
30180309 500 mg
30180326 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30180250 Leverandør Toronto Research Chemicals Leverandørnr. TRCE548955100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 33280-95-4
Molekylær formel C6H9NO3
Molekylvægt (g/mol) 143.14
Synonym (S)-Ethyl Ester-2-isocyanato-propanoic Acid
IUPAC navn ethyl (2S)-2-isocyanatopropanoate
SMIL CCOC(=O)[C@H](C)N=C=O
Tekniske data
Kemisk navn eller materiale (2S)-2-Ethyl Ester-isocyanato-propanoic Acid
Mængde 100 mg
InChI formel InChI=1S/C6H9NO3/c1-3-10-6(9)5(2)7-4-8/h5H,3H2,1-2H3/t5-/m0/s1
Formel vægt 143.0582