missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
(2S)-2-Amino-benzenebutanoic Acid Benzyl Ester Tosylate Salt (>90%), TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | (2S)-2-Amino-benzenebutanoic Acid Benzyl Ester, Tosylate Sal |
| CAS | 117560-24-4 |
| Molekylær formel | C17 H19 N O2 . C7 H8 O3 S |
| Mængde | 1 g |
| Synonym | (2S)-2-Amino-Benzenebutanoic Acid Benzyl Ester Tosylate Salt |
| InChI formel | InChI=1S/C17H19NO2.C7H8O3S/c18-16(12-11-14-7-3-1-4-8-14)17(19)20-13-15-9-5-2-6-10-15;1-6-2-4-7(5-3-6)11(8,9)10/h1-10,16H,11-13,18H2;2-5H,1H3,(H,8,9,10)/t16-;/m0./s1 |
| SMIL | Cc1ccc(cc1)S(=O)(=O)O.N[C@@H](CCc2ccccc2)C(=O)OCc3ccccc3 |
| IUPAC navn | benzyl (2S)-2-amino-4-phenylbutanoate;4-methylbenzenesulfonic acid |
| Molekylvægt (g/mol) | 441.54 |
| Formel vægt | 441.161 |
Produkttitel
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?