Få mere at vide
(2S)-2-Amino-1-phenylpropan-1-one Hydrochloride (Cathinone Hydrochloride), Mikromol
Artikelnummer. 30458174
Skift visning
Mængde:
25 mg
Artikelnummer. Mængde
30458174 25 mg
1 options
Artikelnummer. 30458174 Leverandør Mikromol Leverandørnr. MM1360.020025
Denne vare kræver en prisopdatering, inden den kan bestilles.
Anmod om en pris på denne vare her.
Discover Mikromol- your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
(2S)-2-Amino-1-phenylpropan-1-one Hydrochloride (Cathinone Hydrochloride), Mikromol
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale (-)-(S)-Cathinone hydrochloride
CAS 72739-14-1
Analyt- eller komponentnavne (-)-(S)-Cathinone hydrochloride
Forsendelsestilstand Room Temperature
Molekylær formel C9 H11 N O . Cl H
Certificeringer/compliance ISO 17025
Synonym (S)-(-)-Cathinone Hydrochloride, (2S)-2-Amino-1-phenyl-1-propanone hydrochloride, (S)-2-Amino-1-phenyl-1-propanone hydrochloride, (-)-(S)-Cathinone hydrochloride, (-)-Cathinone hydrochloride, Amfetamine Sulfate Impurity C as Hydrochloride, Dexamfetamine Sulfate Impurity C as Hydrochloride, Phenylpropanolamine Hydrochloride Impurity B as (S)-Enantiomer Hydrochloride, Amfetamine Impurity C as Hydrochloride, Dexamfetamine Impurity C as Hydrochloride, Phenylpropanolamine Impurity B as (S)-Enantiomer Hydrochloride
InChI formel InChI=1S/C9H11NO.ClH/c1-7(10)9(11)8-5-3-2-4-6-8;/h2-7H,10H2,1H3;1H/t7-;/m0./s1
Løsningstype Neat
SMIL Cl.C[C@H](N)C(=O)c1ccccc1
IUPAC navn (2S)-2-amino-1-phenylpropan-1-one;hydrochloride
Molekylvægt (g/mol) 185.65
Mængde 25 mg
Formel vægt 185.0607 g/mol