Få mere at vide
(2S)-2-Acetamido-5-(diaminomethylideneamino)pentanoic Acid, TRC
Artikelnummer. 30415686
Skift visning
Mængde:
2.5 g
5 g
Pakningsstørrelse:
2.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30415686 5 g -
30412922 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30415686 Leverandør Toronto Research Chemicals Leverandørnr. TRCS8083585G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale N-Acetyl-L-arginine
CAS 155-84-0
Molekylær formel C8 H16 N4 O3
Mængde 5 g
Synonym N2-Acetyl-L-arginine
InChI formel InChI=1S/C8H16N4O3/c1-5(13)12-6(7(14)15)3-2-4-11-8(9)10/h6H,2-4H2,1H3,(H,12,13)(H,14,15)(H4,9,10,11)/t6-/m0/s1
SMIL CC(=O)N[C@@H](CCCNC(=N)N)C(=O)O
IUPAC navn (2S)-2-acetamido-5-carbamimidamidopentanoic acid
Molekylvægt (g/mol) 216.2376
Formel vægt 216.1222