Få mere at vide
(2S)-2-[[(1S)-1-(Ethoxycarbonyl)-3-phenylpropyl]amino]propanoic Acid Ethyl Ester Hydrochloride, Mikromol™
Artikelnummer. 30052725
Skift visning
Mængde:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30052725 100 mg
30049121 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30052725 Leverandør Mikromol Leverandørnr. MM0010.15
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
(2S)-2-[[(1S)-1-(Ethoxycarbonyl)-3-phenylpropyl]amino]propanoic Acid Ethyl Ester Hydrochloride
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale (2S)-2-[((1S)-1-(Ethoxycarbonyl)-3-phenylpropyl)amino]propanoic Acid Ethyl Ester Hydrochloride
CAS 122375-72-8
Analyt- eller komponentnavne (2S)-2-[((1S)-1-(Ethoxycarbonyl)-3-phenylpropyl)amino]propanoic Acid Ethyl Ester Hydrochloride
Forsendelsestilstand Room Temperature
Molekylær formel C17 H25 N O4 . Cl H
Certificeringer/compliance ISO 9001
Synonym (2S)-2-[[(1S)-1-(Ethoxycarbonyl)-3-phenylpropyl]amino]propanoic Acid Ethyl Ester Hydrochloride
InChI formel InChI=1S/C17H25NO4.ClH/c1-4-21-16(19)13(3)18-15(17(20)22-5-2)12-11-14-9-7-6-8-10-14;/h6-10,13,15,18H,4-5,11-12H2,1-3H3;1H/t13-,15-;/m0./s1
SMIL Cl.CCOC(=O)[C@H](C)N[C@@H](CCc1ccccc1)C(=O)OCC
IUPAC navn ethyl (2S)-2-[[(2S)-1-ethoxy-1-oxopropan-2-yl]amino]-4-phenylbutanoate;hydrochloride
Molekylvægt (g/mol) 343.85
Mængde 100 mg
Formel vægt 343.155 g/mol