Få mere at vide
(2S)-1-(2-Chloroacetyl)-2-9-pyrrolidinecarbonitrile, TRC
Artikelnummer. 30012741
Skift visning
Mængde:
100 mg
250 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30012741 250 mg
30012502 100 mg
30012551 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30012741 Leverandør Toronto Research Chemicals Leverandørnr. TRCC363780250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 207557-35-5
Molekylær formel C7 H9 Cl N2 O
Molekylvægt (g/mol) 172.61
Synonym 2-Pyrrolidinecarbonitrile, 1-(2-chloroacetyl)-, (2S)-, 2-Pyrrolidinecarbonitrile, 1-(chloroacetyl)-, (2S)- (9CI), (2S)-1-(2-Chloroacetyl)-2-pyrrolidinecarbonitrile, (2S)-1-(Chloroacetyl)-2-pyrrolidinecarbonitrile, (2S)-1-chloroacetylpyrrolidine-2-carbonitrile, (2S)-N-Chloroacetyl-2-cyanopyrrolidine, (S)-1-(2-Chloroacetyl)pyrrolidine-2-carbonitrile, (S)-1-(Chloroacetyl)pyrrolidine-2-carbonitrile, 1-(2-Chloroacetyl)-(2S)-pyrrolidine carbonitrile, 1-Chloroacetyl-2-(S)-cyanopyrrolidine
IUPAC navn (2S)-1-(2-chloroacetyl)pyrrolidine-2-carbonitrile
SMIL ClCC(=O)N1CCC[C@H]1C#N
Tekniske data
Kemisk navn eller materiale (2S)-1-(2-Chloroacetyl)pyrrolidine-2-carbonitrile
Renhedsgrad noter HPLC
Mængde 250 mg
InChI formel InChI=1S/C7H9ClN2O/c8-4-7(11)10-3-1-2-6(10)5-9/h6H,1-4H2/t6-/m0/s1
Formel vægt 172.0403
Procent renhed >95%