Få mere at vide
(2RS,4S)-5,5-Dimethyl-2-[[(phenoxyacetyl)amino]methyl]thiazolidine-4-carboxylic Acid (Penilloic Acids of Phenoxymethylpenicillin), Mikromol™
Artikelnummer. 30050015
Skift visning
Mængde:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30050015 25 mg
30049810 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30050015 Leverandør Mikromol Leverandørnr. MM0026.050025
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
(2RS,4S)-5,5-Dimethyl-2-[[(phenoxyacetyl)amino]methyl]thiazolidine-4-carboxylic Acid (Penilloic Acids of Phenoxymethylpenicillin)
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Penilloic Acids of Phenoxymethylpenicillin
CAS 2714805-53-3
Analyt- eller komponentnavne Penilloic Acids of Phenoxymethylpenicillin
Forsendelsestilstand Room Temperature
Molekylær formel C15 H20 N2 O4 S
Certificeringer/compliance ISO 17025
Synonym Phenoxymethylpenicillin Potassium Imp. F (EP), Phenoxymethylpenicillin Imp. F (EP), (2RS,4S)-5,5-Dimethyl-2-[[(phenoxyacetyl)amino]methyl]thiazolidine-4-carboxylic Acid, Penilloic Acids of Phenoxymethylpenicillin, Phenoxymethylpenicillin Potassium Impurity F, Phenoxymethylpenicillin Impurity F
InChI formel InChI=1S/C22H28N2O2.ClH/c1-2-22(25)23(20-11-7-4-8-12-20)21-14-17-24(26,18-15-21)16-13-19-9-5-3-6-10-19;/h3-12,21H,2,13-18H2,1H3;1H
SMIL CC1(C)SC(CNC(=O)COc2ccccc2)N[C@H]1C(=O)O
IUPAC navn (4S)-5,5-dimethyl-2-[[(2-phenoxyacetyl)amino]methyl]-1,3-thiazolidine-4-carboxylic acid
Molekylvægt (g/mol) 324.4
Mængde 25 mg
Formel vægt 324.1144 g/mol