Få mere at vide
(2RS,4S)-2-[[(Phenylacetyl)amino]methyl]-5,5-dimethylthiazolidine-4-carboxylic Acid (Penilloic Acids of Benzylpenicillin), Mikromol™
Beskrivelse
(2RS,4S)-2-[[(Phenylacetyl)amino]methyl]-5,5-dimethylthiazolidine-4-carboxylic Acid (Penilloic Acids of Benzylpenicillin)
Tekniske data
Tekniske data
| Kemisk navn eller materiale | (2RS,4S)-2-([(Phenylacetyl)amino]methyl)-5,5-dimethylthiazolidine-4-carboxylic Acid (Penilloic Acids of Benzylpenicillin) |
| CAS | 501-34-8 |
| Analyt- eller komponentnavne | (2RS,4S)-2-([(Phenylacetyl)amino]methyl)-5,5-dimethylthiazolidine-4-carboxylic Acid (Penilloic Acids of Benzylpenicillin) |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C15 H20 N2 O3 S |
| Certificeringer/compliance | ISO 17025 |
| Synonym | Benzylpenicillin Procaine Imp. C (EP), Benzylpenicillin Potassium Imp. F (EP), Benzylpenicillin Benzathine Imp. F (EP), (2RS,4S)-2-[[(Phenylacetyl)amino]methyl]-5,5-dimethylthiazolidine-4-carboxylic Acid, Penilloic Acids of Benzylpenicillin, Benzylpenicillin Benzathine Impurity F, Benzylpenicillin Potassium Impurity F, Benzylpenicillin Procaine Impurity C, Carbenicillin Sodium Impurity H, Carbenicillin Impurity H, Benzylpenicillin Impurity F |
| InChI formel | InChI=1S/C15H20N2O3S/c1-15(2)13(14(19)20)17-12(21-15)9-16-11(18)8-10-6-4-3-5-7-10/h3-7,12-13,17H,8-9H2,1-2H3,(H,16,18)(H,19,20)/t12?,13-/m0/s1 |
| SMIL | CC1(C)SC(CNC(=O)Cc2ccccc2)N[C@H]1C(=O)O |
| IUPAC navn | (4S)-5,5-dimethyl-2-[[(2-phenylacetyl)amino]methyl]-1,3-thiazolidine-4-carboxylic acid |
| Vis mere |