Få mere at vide
(2RS)-4-[(1-Methylethyl)amino]-2-phenyl-2-(pyridin-2-yl)butanamide Hydrogen Oxalate, Mikromol™
Artikelnummer. 30049425
Skift visning
Mængde:
25 mg
Artikelnummer. Mængde
30049425 25 mg
1 options
Artikelnummer. 30049425 Leverandør Mikromol Leverandørnr. MM0108.040025
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
(2RS)-4-[(1-Methylethyl)amino]-2-phenyl-2-(pyridin-2-yl)butanamide Hydrogen Oxalate
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale (2RS)-4-[(1-Methylethyl)amino]-2-phenyl-2-(pyridin-2-yl)butanamide Hydrogen Oxalate
CAS 88269-33-4
Analyt- eller komponentnavne (2RS)-4-[(1-Methylethyl)amino]-2-phenyl-2-(pyridin-2-yl)butanamide Hydrogen Oxalate
Forsendelsestilstand Room Temperature
Molekylær formel C18 H23 N3 O . x[C2 H2 O4]
Certificeringer/compliance ISO 9001
Synonym (2RS)-4-[(1-Methylethyl)amino]-2-phenyl-2-(pyridin-2-yl)butanamide Hydrogen Oxalate, (+/-)-alpha-[2-[(1-Methylethyl)amino]ethyl]-alpha-phenyl-2-pyridineacetamide ethanedioate, Disopyramide Impurity C as Hydrogen Oxalate
InChI formel InChI=1S/C18H23N3O.C2H2O4/c1-14(2)20-13-11-18(17(19)22,15-8-4-3-5-9-15)16-10-6-7-12-21-16;3-1(4)2(5)6/h3-10,12,14,20H,11,13H2,1-2H3,(H2,19,22);(H,3,4)(H,5,6)
SMIL CC(C)NCCC(C(=O)N)(c1ccccc1)c2ccccn2.OC(=O)C(=O)O
IUPAC navn oxalic acid;2-phenyl-4-(propan-2-ylamino)-2-pyridin-2-ylbutanamide