Få mere at vide
(2RS)-3-(Dimethylamino)-2-methyl-1-phenylpropan-1-one Hydrochloride, Mikromol™
Beskrivelse
(2RS)-3-(Dimethylamino)-2-methyl-1-phenylpropan-1-one Hydrochloride
Tekniske data
Tekniske data
| Kemisk navn eller materiale | (2RS)-3-(Dimethylamino)-2-methyl-1-phenylpropan-1-one Hydrochloride |
| CAS | 5400-92-0 |
| Analyt- eller komponentnavne | (2RS)-3-(Dimethylamino)-2-methyl-1-phenylpropan-1-one Hydrochloride |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C12 H17 N O . Cl H |
| Certificeringer/compliance | ISO/IEC 17025 |
| Synonym | Dextropropoxyphene Hydrochloride Imp. F (EP), Dextropropoxyphene Imp. F (EP), (2RS)-3-(Dimethylamino)-2-methyl-1-phenylpropan-1-one Hydrochloride, Dextropropoxyphene Hydrochloride Imp. F (EP) as Hydrochloride, Dextropropoxyphene Hydrochloride Impurity F as Hydrochloride, Dextropropoxyphene Impurity F as Hydrochloride |
| InChI formel | InChI=1S/C12H17NO.ClH/c1-10(9-13(2)3)12(14)11-7-5-4-6-8-11;/h4-8,10H,9H2,1-3H3;1H |
| SMIL | Cl.CC(CN(C)C)C(=O)c1ccccc1 |
| IUPAC navn | 3-(dimethylamino)-2-methyl-1-phenylpropan-1-one;hydrochloride |
| Vis mere |