Få mere at vide
(2RS)-3-(5H-Dibenzo[b,f]azepin-5-yl)-N,N,2-trimethylpropan-1-amine Hydrochloride, Mikromol™
Beskrivelse
(2RS)-3-(5H-Dibenzo[b,f]azepin-5-yl)-N,N,2-trimethylpropan-1-amine Hydrochloride
Tekniske data
Tekniske data
| Kemisk navn eller materiale | (2RS)-3-(5H-Dibenzo[b,f]azepin-5-yl)-N,N,2-trimethylpropan-1-amine Hydrochloride |
| CAS | 1159-80-4 |
| Analyt- eller komponentnavne | (2RS)-3-(5H-Dibenzo[b,f]azepin-5-yl)-N,N,2-trimethylpropan-1-amine Hydrochloride |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C20 H24 N2 . Cl H |
| Certificeringer/compliance | ISO 9001 |
| Synonym | Trimipramine Maleate Imp. C (EP), Trimipramine Imp. C (EP), Trimipramine USP Related Compound A, Trimipramine USP RC A, (2RS)-3-(5H-Dibenzo[b,f]azepin-5-yl)-N,N,2-trimethylpropan-1-amine Hydrochloride, Trimipramine Maleate Imp. C (EP) as Hydrochloride, Trimipramine Related Compound A as Hydrochloride, Trimipramine Maleate Impurity C as Hydrochloride, Trimipramine Impurity C as Hydrochloride |
| InChI formel | InChI=1S/C20H24N2.ClH/c1-16(14-21(2)3)15-22-19-10-6-4-8-17(19)12-13-18-9-5-7-11-20(18)22;/h4-13,16H,14-15H2,1-3H3;1H |
| SMIL | Cl.CC(CN(C)C)CN1c2ccccc2C=Cc3ccccc13 |
| IUPAC navn | 3-benzo[b][1]benzazepin-11-yl-N,N,2-trimethylpropan-1-amine;hydrochloride |
| Vis mere |