Få mere at vide
(2RS)-2-(Dimethylamino)-2-phenylbutanol Hydrochloride, Mikromol™
Artikelnummer. 30463178
Skift visning
Mængde:
25 mg
100 mg
4 x 25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30463178 4 x 25 mg
17974296 25 mg
17902346 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30463178 Leverandør Mikromol Leverandørnr. MM0417.0100254AMP
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
(2RS)-2-(Dimethylamino)-2-phenylbutanol Hydrochloride, Mikromol
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale (2RS)-2-(Dimethylamino)-2-phenylbutanol Hydrochloride
CAS 60577-23-3
Analyt- eller komponentnavne (2RS)-2-(Dimethylamino)-2-phenylbutanol Hydrochloride
Forsendelsestilstand Room Temperature
Molekylær formel C12 H19 N O . Cl H
Certificeringer/compliance ISO 17025
Synonym Trimebutine Imp. A (EP), (2RS)-2-(Dimethylamino)-2-phenylbutanol Hydrochloride, Trimebutine Imp. A (EP) as Hydrochloride, Trimebutine Maleate Impurity A as Hydrochloride, Trimebutine Impurity A as Hydrochloride
InChI formel InChI=1S/C12H19NO.ClH/c1-4-12(10-14,13(2)3)11-8-6-5-7-9-11;/h5-9,14H,4,10H2,1-3H3;1H
Løsningstype Neat
SMIL Cl.CCC(CO)(N(C)C)c1ccccc1
IUPAC navn 2-(dimethylamino)-2-phenylbutan-1-ol;hydrochloride
Molekylvægt (g/mol) 229.75
Mængde 4 x 25 mg
Formel vægt 229.1233 g/mol