Få mere at vide
(2RS)-2-(Cyclohex-1-enyl)-2-(4-methoxyphenyl)-N,N-dimethylethanamine Hydrochloride, Mikromol™
Artikelnummer. 17992936
Skift visning
Mængde:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
17992936 25 mg
17991386 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 17992936 Leverandør Mikromol Leverandørnr. MM0393.100025
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
(2RS)-2-(Cyclohex-1-enyl)-2-(4-methoxyphenyl)-N,N-dimethylethanamine Hydrochloride
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale (2RS)-2-(Cyclohex-1-enyl)-2-(4-methoxyphenyl)-N,N-dimethylethanamine Hydrochloride
CAS 93413-79-7
Analyt- eller komponentnavne (2RS)-2-(Cyclohex-1-enyl)-2-(4-methoxyphenyl)-N,N-dimethylethanamine Hydrochloride
Forsendelsestilstand Room Temperature
Molekylær formel C17 H25 N O . Cl H
Certificeringer/compliance ISO 17025
Synonym Venlafaxine Hydrochloride Imp. F (EP), Venlafaxine Imp. F (EP), (2RS)-2-(Cyclohex-1-enyl)-2-(4-methoxyphenyl)-N,N-dimethylethanamine Hydrochloride, Venlafaxine Hydrochloride Imp. F (EP) as Hydrochloride, Venlafaxine Hydrochloride Impurity F as Hydrochloride, Venlafaxine Impurity F as Hydrochloride
InChI formel InChI=1S/C17H25NO.ClH/c1-18(2)13-17(14-7-5-4-6-8-14)15-9-11-16(19-3)12-10-15;/h7,9-12,17H,4-6,8,13H2,1-3H3;1H
SMIL Cl.COc1ccc(cc1)C(CN(C)C)C2=CCCCC2
IUPAC navn 2-(cyclohexen-1-yl)-2-(4-methoxyphenyl)-N,N-dimethylethanamine;hydrochloride
Molekylvægt (g/mol) 295.85
Mængde 25 mg
Formel vægt 295.1703 g/mol