Få mere at vide
(2RS)-2-Benzyl-4-(dimethylamino)butanoic Acid Hydrochloride, Mikromol™
Beskrivelse
(2RS)-2-Benzyl-4-(dimethylamino)butanoic Acid Hydrochloride
Tekniske data
Tekniske data
| Kemisk navn eller materiale | (2RS)-2-Benzyl-4-(dimethylamino)butanoic Acid Hydrochloride |
| CAS | 1823817-92-0 |
| Analyt- eller komponentnavne | (2RS)-2-Benzyl-4-(dimethylamino)butanoic Acid Hydrochloride |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C13 H19 N O2 . Cl H |
| Certificeringer/compliance | ISO 9001 |
| Synonym | Dimetindene Maleate Imp. D (EP), Dimetindene Imp. D (EP), (2RS)-2-Benzyl-4-(dimethylamino)butanoic Acid Hydrochloride, Dimetindene Maleate Imp. D (EP) as Hydrochloride, Dimetindene Maleate Impurity D as Hydrochloride, Dimetindene Impurity D as Hydrochloride |
| InChI formel | InChI=1S/C19H21ClN2O2.2ClH/c20-17-8-6-16(7-9-17)19(15-4-2-1-3-5-15)22-12-10-21(11-13-22)14-18(23)24;;/h1-9,19H,10-14H2,(H,23,24);2*1H |
| SMIL | Cl.CN(C)CCC(Cc1ccccc1)C(=O)O |
| IUPAC navn | 2-benzyl-4-(dimethylamino)butanoic acid;hydrochloride |
| Vis mere |