Få mere at vide
(2RS)-2-Amino-3-hydroxypropanohydrazide Hydrochloride, Mikromol™
Beskrivelse
(2RS)-2-Amino-3-hydroxypropanohydrazide Hydrochloride
Tekniske data
Tekniske data
| Kemisk navn eller materiale | (2RS)-2-Amino-3-hydroxypropanohydrazide Hydrochloride |
| CAS | 55819-71-1 |
| Analyt- eller komponentnavne | (2RS)-2-Amino-3-hydroxypropanohydrazide Hydrochloride |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C3 H9 N3 O2 . Cl H |
| Certificeringer/compliance | ISO 9001 |
| Synonym | Serine, hydrazide, monohydrochloride (9CI), DL-Serine, hydrazide, monohydrochloride, DL-Serine hydrazide hydrochloride, Benserazide Hydrochloride Impurity A as Hydrochloride, Benserazide Impurity A as Hydrochloride |
| InChI formel | InChI=1S/C3H9N3O2.ClH/c4-2(1-7)3(8)6-5;/h2,7H,1,4-5H2,(H,6,8);1H |
| SMIL | Cl.NNC(=O)C(N)CO |
| IUPAC navn | 2-amino-3-hydroxypropanehydrazide;hydrochloride |
| Vis mere |