Få mere at vide
(2RS)-2-(4-Formylphenyl)propanoic Acid, Mikromol™
Artikelnummer. 30050671
Skift visning
Mængde:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30050671 25 mg
17951056 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30050671 Leverandør Mikromol Leverandørnr. MM0002.260025
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
(2RS)-2-(4-Formylphenyl)propanoic Acid
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale (2RS)-2-(4-Formylphenyl)propanoic Acid
CAS 43153-07-7
Analyt- eller komponentnavne (2RS)-2-(4-Formylphenyl)propanoic Acid
Forsendelsestilstand Ice pack (-20°C)
Molekylær formel C10 H10 O3
Certificeringer/compliance ISO 17025
Synonym (2RS)-2-(4-Formylphenyl)propanoic Acid, Ibuprofen Imp. K (EP), Ibuprofen Impurity K
InChI formel InChI=1S/C10H10O3/c1-7(10(12)13)9-4-2-8(6-11)3-5-9/h2-7H,1H3,(H,12,13)
SMIL CC(C(=O)O)c1ccc(C=O)cc1
IUPAC navn 2-(4-formylphenyl)propanoic acid
Molekylvægt (g/mol) 178.18
Mængde 25 mg
Formel vægt 178.063 g/mol