Få mere at vide
(2RS)-2-[(4-Chlorobenzyl)oxy]-2-(2,4-dichlorophenyl)ethanamine Nitrate, Mikromol™
Artikelnummer. 17993916
Skift visning
Mængde:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
17993916 100 mg
17920716 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 17993916 Leverandør Mikromol Leverandørnr. MM0524.02
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
(2RS)-2-[(4-Chlorobenzyl)oxy]-2-(2,4-dichlorophenyl)ethanamine Nitrate
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2-[(4-Chlorobenzyl)oxy]-2-(2,4-dichlorophenyl)ethanamine nitrate
CAS 2724510-72-7
Analyt- eller komponentnavne 2-[(4-Chlorobenzyl)oxy]-2-(2,4-dichlorophenyl)ethanamine nitrate
Forsendelsestilstand Room Temperature
Molekylær formel C15 H14 Cl3 N O . H N O3
Certificeringer/compliance ISO 17025
Synonym Econazole Nitrate Imp. B (EP), Econazole USP Related Compound B, Econazole USP RC B, (2RS)-2-[(4-Chlorobenzyl)oxy]-2-(2,4-dichlorophenyl)ethanamine Nitrate, Econazole Nitrate Imp. B (EP) as Nitrate, Econazole Related Compound B as Nitrate, Econazole Nitrate Impurity B as Nitrate, Econazole Impurity B as Nitrate
InChI formel InChI=1S/C15H14Cl3NO.HNO3/c16-11-3-1-10(2-4-11)9-20-15(8-19)13-6-5-12(17)7-14(13)18;2-1(3)4/h1-7,15H,8-9,19H2;(H,2,3,4)
SMIL NCC(OCc1ccc(Cl)cc1)c2ccc(Cl)cc2Cl.O[N+](=O)[O-]
IUPAC navn 2-[(4-chlorophenyl)methoxy]-2-(2,4-dichlorophenyl)ethanamine;nitric acid
Molekylvægt (g/mol) 393.65
Mængde 100 mg
Formel vægt 392.0097 g/mol