Få mere at vide
(2RS)-2-[(4-Chlorobenzyl)oxy]-2-(2,4-dichlorophenyl)ethanamine Nitrate, Mikromol™
Beskrivelse
(2RS)-2-[(4-Chlorobenzyl)oxy]-2-(2,4-dichlorophenyl)ethanamine Nitrate
Tekniske data
Tekniske data
| Kemisk navn eller materiale | 2-[(4-Chlorobenzyl)oxy]-2-(2,4-dichlorophenyl)ethanamine nitrate |
| CAS | 2724510-72-7 |
| Analyt- eller komponentnavne | 2-[(4-Chlorobenzyl)oxy]-2-(2,4-dichlorophenyl)ethanamine nitrate |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C15 H14 Cl3 N O . H N O3 |
| Certificeringer/compliance | ISO 17025 |
| Synonym | Econazole Nitrate Imp. B (EP), Econazole USP Related Compound B, Econazole USP RC B, (2RS)-2-[(4-Chlorobenzyl)oxy]-2-(2,4-dichlorophenyl)ethanamine Nitrate, Econazole Nitrate Imp. B (EP) as Nitrate, Econazole Related Compound B as Nitrate, Econazole Nitrate Impurity B as Nitrate, Econazole Impurity B as Nitrate |
| InChI formel | InChI=1S/C15H14Cl3NO.HNO3/c16-11-3-1-10(2-4-11)9-20-15(8-19)13-6-5-12(17)7-14(13)18;2-1(3)4/h1-7,15H,8-9,19H2;(H,2,3,4) |
| SMIL | NCC(OCc1ccc(Cl)cc1)c2ccc(Cl)cc2Cl.O[N+](=O)[O-] |
| IUPAC navn | 2-[(4-chlorophenyl)methoxy]-2-(2,4-dichlorophenyl)ethanamine;nitric acid |
| Vis mere |