missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(2RS)-2-(4-Chloro-phenyl)-4-(dimethylamino)-2-(pyridin-2-yl)butanenitrile Maleate, Mikromol™
Description
(2RS)-2-(4-Chloro-phenyl)-4-(dimethylamino)-2-(pyridin-2-yl)butanenitrile Maleate
Specifications
Specifications
| Kemisk navn eller materiale | (2RS)-2-(4-Chloro-phenyl)-4-(dimethylamino)-2-(pyridin-2-yl)butanenitrile Maleate |
| CAS | 2995356-12-0 |
| Analyt- eller komponentnavne | (2RS)-2-(4-Chloro-phenyl)-4-(dimethylamino)-2-(pyridin-2-yl)butanenitrile Maleate |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C17 H18 Cl N3 . C4 H4 O4 |
| Certificeringer/compliance | ISO 17025 |
| Synonym | Chlorphenamine Imp. D (EP), (2RS)-2-(4-Chloro-phenyl)-4-(dimethylamino)-2-(pyridin-2-yl)butanenitrile Maleate, Chlorphenamine Maleate Imp. D (EP) as Maleate |
| InChI formel | InChI=1S/C17H20N2S.ClH/c1-13(12-18(2)3)19-14-8-4-6-10-16(14)20-17-11-7-5-9-15(17)19;/h4-11,13H,12H2,1-3H3;1H |
| SMIL | CN(C)CCC(C#N)(c1ccc(Cl)cc1)c2ccccn2.OC(=O)\C=C/C(=O)O |
| IUPAC navn | (Z)-but-2-enedioic acid;2-(4-chlorophenyl)-4-(dimethylamino)-2-pyridin-2-ylbutanenitrile |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?