Få mere at vide
(2RS)-2-(3-Benzoylphenyl)propanenitrile, Mikromol™
Artikelnummer. 17982066
Skift visning
Mængde:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
17982066 25 mg
17980826 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 17982066 Leverandør Mikromol Leverandørnr. MM0001.040025
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
(2RS)-2-(3-Benzoylphenyl)propanenitrile
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale (2RS)-2-(3-Benzoylphenyl)propanenitrile
CAS 42872-30-0
Analyt- eller komponentnavne (2RS)-2-(3-Benzoylphenyl)propanenitrile
Forsendelsestilstand Room Temperature
Molekylær formel C16 H13 N O
Certificeringer/compliance ISO 17025
Synonym Ketoprofen Imp. F (EP), Ketoprofen USP Related Compound F, Ketoprofen USP RC F, (2RS)-2-(3-Benzoylphenyl)propanenitrile, Ketoprofen Related Compound F, Ketoprofen Impurity F
InChI formel InChI=1S/C16H13NO/c1-12(11-17)14-8-5-9-15(10-14)16(18)13-6-3-2-4-7-13/h2-10,12H,1H3
SMIL CC(C#N)c1cccc(c1)C(=O)c2ccccc2
IUPAC navn 2-(3-benzoylphenyl)propanenitrile
Molekylvægt (g/mol) 235.28
Mængde 25 mg
Formel vægt 235.0997 g/mol