Få mere at vide
(2RS)-2-(2-Oxopyrrolidin-1-yl)butanoic Acid, Mikromol™
Artikelnummer. 30050329
Skift visning
Mængde:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30050329 100 mg
30050565 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30050329 Leverandør Mikromol Leverandørnr. MM1372.01
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
(2RS)-2-(2-Oxopyrrolidin-1-yl)butanoic Acid
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2-(2-oxopyrrolidin-1-yl)butanoic acid
CAS 67118-31-4
Analyt- eller komponentnavne 2-(2-oxopyrrolidin-1-yl)butanoic acid
Forsendelsestilstand Room Temperature
Molekylær formel C8 H13 N O3
Certificeringer/compliance ISO 17025
Synonym Levetiracetam Imp. A (EP), 1-Pyrrolidineacetic acid, alpha-ethyl-2-oxo-, Levetiracetam Impurity A
InChI formel InChI=1S/C8H13NO3/c1-2-6(8(11)12)9-5-3-4-7(9)10/h6H,2-5H2,1H3,(H,11,12)
SMIL CCC(N1CCCC1=O)C(=O)O
IUPAC navn 2-(2-oxopyrrolidin-1-yl)butanoic acid
Molekylvægt (g/mol) 171.19
Mængde 100 mg
Formel vægt 171.0895 g/mol