Learn More
(2RS)-2-(2,4-Difluorophenyl)-1-(1H-1,2,4-triazol-1-yl)-3-(4H-1,2,4-triazol-4-yl)propan-2-ol, LoGiCal
Product Code. 30458109
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Product Code. Quantity Unit Size
30458109 10 mg 10mg
1 options
Product Code. 30458109 Supplier LoGiCal Supplier No. LGCFOR0235.11
 Restricted Item
Forventet forsendelse: 30-11-2026
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material (2RS)-2-(2,4-Difluorophenyl)-1-(1H-1,2,4-triazol-1-yl)-3-(4H-1,2,4-triazol-4-yl)propan-2-ol
CAS 89429-59-4
Analyte or Component Names (2RS)-2-(2,4-Difluorophenyl)-1-(1H-1,2,4-triazol-1-yl)-3-(4H-1,2,4-triazol-4-yl)propan-2-ol
Shipping Condition Room Temperature
Molecular Formula C13 H12 F2 N6 O
Certifications/Compliance ISO 9001
Synonym Fluconazole Imp. A (EP), (2RS)-2-(2,4-Difluorophenyl)-1-(1H-1,2,4-triazol-1-yl)-3-(4H-1,2,4-triazol-4-yl)propan-2-ol, Fluconazole Impurity A
InChI Formula InChI=1S/C13H12F2N6O/c14-10-1-2-11(12(15)3-10)13(22,4-20-8-17-18-9-20)5-21-7-16-6-19-21/h1-3,6-9,22H,4-5H2
Solution Type Neat
SMILES OC(Cn1cnnc1)(Cn2cncn2)c3ccc(F)cc3F
IUPAC Name 2-(2,4-difluorophenyl)-1-(1,2,4-triazol-1-yl)-3-(1,2,4-triazol-4-yl)propan-2-ol
Industry Type Food Testing
Molecular Weight (g/mol) 306.27
Quantity 10 mg
Formula Weight 306.1041 g/mol