Få mere at vide
(2RS)-1-(Isopropylamino)-3-[4-(2-methoxyethoxy)methylphenoxy]propan-2-ol, Mikromol™
Artikelnummer. 17941256
Skift visning
Mængde:
25 mg
100 mg
4 x 25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
17941256 100 mg
17993856 25 mg
30460890 4 x 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 17941256 Leverandør Mikromol Leverandørnr. MM0460.05
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
(2RS)-1-(Isopropylamino)-3-[4-(2-methoxyethoxy)methylphenoxy]propan-2-ol
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale (2RS)-1-(Isopropylamino)-3-[4-(2-methoxyethoxy)methylphenoxy]propan-2-ol
CAS 1346604-00-9
Analyt- eller komponentnavne (2RS)-1-(Isopropylamino)-3-[4-(2-methoxyethoxy)methylphenoxy]propan-2-ol
Forsendelsestilstand Room Temperature
Molekylær formel C16 H27 N O4
Certificeringer/compliance ISO 17025
Synonym Bisoprolol Fumarate Imp. Q (EP), Bisoprolol Imp. Q (EP), (2RS)-1-(Isopropylamino)-3-[4-(2-methoxyethoxy)methylphenoxy]propan-2-ol, Bisoprolol Fumarate Impurity Q, Bisoprolol Impurity Q
InChI formel InChI=1S/C9H11FN2O5/c1-3-5(13)6(14)8(17-3)12-2-4(10)7(15)11-9(12)16/h2-3,5-6,8,13-14H,1H3,(H,11,15,16)/t3-,5-,6-,8-/m1/s5
SMIL COCCOCc1ccc(OCC(O)CNC(C)C)cc1
IUPAC navn 1-[4-(2-methoxyethoxymethyl)phenoxy]-3-(propan-2-ylamino)propan-2-ol
Molekylvægt (g/mol) 297.39
Mængde 100 mg
Formel vægt 297.194 g/mol