Få mere at vide
(2RS)-1-Bromo-2-(2,4-difluorophenyl)-3-(1H-1,2,4-triazol-1-yl)propan-2-ol, Mikromol™
Artikelnummer. 17920956
Skift visning
Mængde:
25 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
17920956 50 mg
17924036 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 17920956 Leverandør Mikromol Leverandørnr. MM0235.07
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
(2RS)-1-Bromo-2-(2,4-difluorophenyl)-3-(1H-1,2,4-triazol-1-yl)propan-2-ol
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale (2RS)-1-Bromo-2-(2,4-difluorophenyl)-3-(1H-1,2,4-triazol-1-yl)propan-2-ol
CAS 150194-52-8
Analyt- eller komponentnavne (2RS)-1-Bromo-2-(2,4-difluorophenyl)-3-(1H-1,2,4-triazol-1-yl)propan-2-ol
Forsendelsestilstand Room Temperature
Molekylær formel C11 H10 Br F2 N3 O
Certificeringer/compliance ISO 17025
Synonym (2RS)-1-Bromo-2-(2,4-difluorophenyl)-3-(1H-1,2,4-triazol-1-yl)propan-2-ol, Fluconazole Imp. H (EP), Fluconazole Impurity H
InChI formel InChI=1S/C11H10BrF2N3O/c12-4-11(18,5-17-7-15-6-16-17)9-2-1-8(13)3-10(9)14/h1-3,6-7,18H,4-5H2
SMIL OC(CBr)(Cn1cncn1)c2ccc(F)cc2F
IUPAC navn 1-bromo-2-(2,4-difluorophenyl)-3-(1,2,4-triazol-1-yl)propan-2-ol
Molekylvægt (g/mol) 318.12
Mængde 50 mg
Formel vægt 316.9975 g/mol