Få mere at vide
(2RS)-1-(4-Ethylphenoxy)-3-[(1-methylethyl)amino]propan-2-ol Hydrochloride, Mikromol™
Beskrivelse
(2RS)-1-(4-Ethylphenoxy)-3-[(1-methylethyl)amino]propan-2-ol Hydrochloride
Tekniske data
Tekniske data
| Kemisk navn eller materiale | (2RS)-1-(4-Ethylphenoxy)-3-[(1-methylethyl)amino]propan-2-ol Hydrochloride |
| CAS | 464877-45-0 |
| Analyt- eller komponentnavne | (2RS)-1-(4-Ethylphenoxy)-3-[(1-methylethyl)amino]propan-2-ol Hydrochloride |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C14 H23 N O2 . Cl H |
| Certificeringer/compliance | ISO 17025 |
| Synonym | Betaxolol Hydrochloride Imp. A (EP), Betaxolol Imp. A (EP), (2RS)-1-(4-Ethylphenoxy)-3-[(1-methylethyl)amino]propan-2-ol Hydrochloride, 2-Propanol, 1-(4-ethylphenoxy)-3-[(1-methylethyl)amino]-, hydrochloride (1:1), Betaxolol Hydrochloride Imp. A (EP) as Hydrochloride, Betaxolol Hydrochloride Impurity A as Hydrochloride, Betaxolol Impurity A as Hydrochloride |
| InChI formel | InChI=1S/C14H23NO2.ClH/c1-4-12-5-7-14(8-6-12)17-10-13(16)9-15-11(2)3;/h5-8,11,13,15-16H,4,9-10H2,1-3H3;1H |
| SMIL | Cl.CCc1ccc(OCC(O)CNC(C)C)cc1 |
| IUPAC navn | 1-(4-ethylphenoxy)-3-(propan-2-ylamino)propan-2-ol;hydrochloride |
| Vis mere |