Få mere at vide
(2RS)-1-(4-Ethenylphenoxy)-3-[(1-methylethyl)amino]propan-2-ol, Mikromol™
Artikelnummer. 30464262
Skift visning
Mængde:
25 mg
100 mg
4 x 25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30464262 4 x 25 mg
30050449 25 mg
30052474 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30464262 Leverandør Mikromol Leverandørnr. MM0027.1100254AMP
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
(2RS)-1-(4-Ethenylphenoxy)-3-[(1-methylethyl)amino]propan-2-ol, Mikromol
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale (2RS)-1-(4-Ethenylphenoxy)-3-[(1-methylethyl)amino]propan-2-ol
CAS 933468-54-3
Analyt- eller komponentnavne (2RS)-1-(4-Ethenylphenoxy)-3-[(1-methylethyl)amino]propan-2-ol
Forsendelsestilstand Room Temperature
Molekylær formel C14 H21 N O2
Certificeringer/compliance ISO 17025
Synonym (2RS)-1-(4-Ethenylphenoxy)-3-[(1-methylethyl)amino]propan-2-ol
InChI formel InChI=1S/C14H21NO2/c1-4-12-5-7-14(8-6-12)17-10-13(16)9-15-11(2)3/h4-8,11,13,15-16H,1,9-10H2,2-3H3
Løsningstype Neat
SMIL CC(C)NCC(O)COc1ccc(C=C)cc1
IUPAC navn 1-(4-ethenylphenoxy)-3-(propan-2-ylamino)propan-2-ol
Molekylvægt (g/mol) 235.32
Mængde 4 x 25 mg
Formel vægt 235.1572 g/mol