Få mere at vide
[(2RS)-1-[4-[(2-Ethoxyethoxy)methyl]phenoxy]-3-isopropylaminopropan-2-ol, Mikromol™
Artikelnummer. 30049540
Skift visning
Mængde:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30049540 100 mg
30049189 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30049540 Leverandør Mikromol Leverandørnr. MM0460.25
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
[(2RS)-1-[4-[(2-Ethoxyethoxy)methyl]phenoxy]-3-isopropylaminopropan-2-ol
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale ((2RS)-1-(4-((2-Ethoxyethoxy)methyl)phenoxy)-3-isopropylaminopropan-2-ol
CAS 1346601-75-9
Analyt- eller komponentnavne ((2RS)-1-(4-((2-Ethoxyethoxy)methyl)phenoxy)-3-isopropylaminopropan-2-ol
Forsendelsestilstand Room Temperature
Molekylær formel C17 H29 N O4
Certificeringer/compliance ISO 17025
Synonym Bisoprolol Fumarate Imp. N (EP), Bisoprolol Imp. N (EP), [(2RS)-1-[4-[(2-Ethoxyethoxy)methyl]phenoxy]-3-isopropylaminopropan-2-ol
InChI formel InChI=1S/C13H19Cl/c1-9-6-11(13(3,4)5)7-10(2)12(9)8-14/h6-7H,8H2,1-5H7
SMIL CCOCCOCc1ccc(OCC(O)CNC(C)C)cc1
IUPAC navn 1-[4-(2-ethoxyethoxymethyl)phenoxy]-3-(propan-2-ylamino)propan-2-ol
Molekylvægt (g/mol) 311.42
Mængde 100 mg
Formel vægt 311.2097 g/mol