Få mere at vide
(2R,6R,7R)- and (2S,6R,7R)-7-[[(2R)-2-Amino-2-phenylacetyl]amino]-3-chloro-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-3-ene-2-carboxylic Acid (Δ-3-Cefaclor), Mikromol™
Artikelnummer. 17971956
Skift visning
Mængde:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
17971956 100 mg
17960506 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 17971956 Leverandør Mikromol Leverandørnr. MM0326.03
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
(2R,6R,7R)- and (2S,6R,7R)-7-[[(2R)-2-Amino-2-phenylacetyl]amino]-3-chloro-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-3-ene-2-carboxylic Acid (Δ-3-Cefaclor)
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Delta-3-Cefaclor
CAS 152575-13-8
Analyt- eller komponentnavne Delta-3-Cefaclor
Forsendelsestilstand Room Temperature
Molekylær formel C15 H14 Cl N3 O4 S
Certificeringer/compliance ISO 17025
Synonym Cefaclor Imp. D (EP), Cefaclor Delta-3 Isomer, 5-Thia-1-azabicyclo[4.2.0]oct-3-ene-2-carboxylic acid, 7-[[(2R)-aminophenylacetyl]amino]-3-chloro-8-oxo-, (6R,7R)- (9CI), 5-Thia-1-azabicyclo[4.2.0]oct-3-ene-2-carboxylic acid, 7-[(aminophenylacetyl)amino]-3-chloro-8-oxo-, [6R-[6alpha,7beta(R*)]]-, Cefaclor Impurity D
InChI formel InChI=1S/C15H14ClN3O4S/c16-8-6-24-14-10(13(21)19(14)11(8)15(22)23)18-12(20)9(17)7-4-2-1-3-5-7/h1-6,9-11,14H,17H2,(H,18,20)(H,22,23)/t9-,10-,11?,14-/m1/s1
SMIL N[C@@H](C(=O)N[C@H]1[C@H]2SC=C(Cl)C(N2C1=O)C(=O)O)c3ccccc3
IUPAC navn (6R,7R)-7-[[(2R)-2-amino-2-phenylacetyl]amino]-3-chloro-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-3-ene-2-carboxylic acid
Molekylvægt (g/mol) 367.81
Mængde 100 mg
Formel vægt 367.0394 g/mol