Få mere at vide
(2R,5S)-L-Menthol-5-(4-amino-2-oxo-1(2H)-pyrimidinyl)-1,3-oxathiolane-2-carboxylate, TRC
Artikelnummer. 30470397
Skift visning
Mængde:
50 mg
250 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30470397 250 mg
30474111 50 mg
30482508 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30470397 Leverandør Toronto Research Chemicals Leverandørnr. TRCM219020250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 147027-10-9
Molekylær formel C18 H27 N3 O4 S
Molekylvægt (g/mol) 381.49
Synonym 1,3-Oxathiolane-2-carboxylic acid, 5-(4-amino-2-oxo-1(2H)-pyrimidinyl)-, (1R,2S,5R)-5-methyl-2-(1-methylethyl)cyclohexyl ester, (2R,5S)-, 1,3-Oxathiolane-2-carboxylic acid, 5-(4-amino-2-oxo-1(2H)-pyrimidinyl)-, 5-methyl-2-(1-methylethyl)cyclohexyl ester, [1R-[1α(2R*,5S*),2β,5α]]-, (2R,5S)-5-(4-Amino-2-oxo-1,2-dihydro-1-pyrimidinyl)-1,3-oxathiolane-2-carboxylic acid (1R,2S,5R)-2-isopropyl-5-methylcyclohexyl ester, GR 146496X, Lamivudine related
IUPAC navn [(1R,2S,5R)-5-methyl-2-propan-2-ylcyclohexyl] (2R,5S)-5-(4-amino-2-oxopyrimidin-1-yl)-1,3-oxathiolane-2-carboxylate
SMIL CC(C)[C@@H]1CC[C@@H](C)C[C@H]1OC(=O)[C@@H]2O[C@@H](CS2)N3C=CC(=NC3=O)N
Tekniske data
Kemisk navn eller materiale (2R,5S)-5-(4-Amino-2-oxo-1,2-dihydro-1-pyrimidinyl)-1,3-oxathiolane-2-carboxylic acid (1R,2S,5R)-2-isopropyl-5-methylcyclohexyl ester
Mængde 250 mg
InChI formel InChI=1S/C18H27N3O4S/c1-10(2)12-5-4-11(3)8-13(12)24-16(22)17-25-15(9-26-17)21-7-6-14(19)20-18(21)23/h6-7,10-13,15,17H,4-5,8-9H2,1-3H3,(H2,19,20,23)/t11-,12+,13-,15+,17-/m1/s1
Formel vægt 381.172