Få mere at vide
(2R,4S)-2-(1,1-Dimethylethyl)-4-methyl-1,3-oxathiolan-5-one, TRC
Artikelnummer. 30375735
Skift visning
Mængde:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30375735 100 mg
30377250 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30375735 Leverandør Toronto Research Chemicals Leverandørnr. TRCD471958100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale (2R,4S)-2-(1,1-Dimethylethyl)-4-methyl-1,3-oxathiolan-5-one
CAS 476371-85-4
Molekylær formel C8H14O2S
Mængde 100 mg
Synonym (2R,4S)-2-(t-Butyl)-4-methyl-1,3-oxathiolan-5-one
InChI formel InChI=1S/C8H14O2S/c1-5-6(9)10-7(11-5)8(2,3)4/h5,7H,1-4H3/t5-,7+/m0/s1
SMIL C[C@@H]1S[C@@H](OC1=O)C(C)(C)C
IUPAC navn (2R,4S)-2-tert-butyl-4-methyl-1,3-oxathiolan-5-one
Molekylvægt (g/mol) 174.26
Formel vægt 174.0715