missing translation for 'onlineSavingsMsg'
Få mere at vide

(2R,4R)1-[(2S)2-Amino-5-[[imino(nitroamino)methyl]amino]-1-oxopentyl]-4-methyl-2-piperidinecarboxylic Acid Ethyl Ester Hydrochloride, TRC

Artikelnummer. 30314625
Change view
Click to view available options
Mængde:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
30314625 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30314625 Supplier Toronto Research Chemicals Supplier No. TRCA61169510MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale 1-[2-Amino-5-[[imino(nitroamino)methyl]amino]-1-oxopentyl]-4
CAS 74874-08-1
Molekylær formel C15 H28 N6 O5 . Cl H
Mængde 10 mg
Synonym Argatroban Related Compound B
InChI formel InChI=1S/C15H28N6O5.ClH/c1-3-26-14(23)12-9-10(2)6-8-20(12)13(22)11(16)5-4-7-18-15(17)19-21(24)25;/h10-12H,3-9,16H2,1-2H3,(H3,17,18,19);1H/t10-,11+,12-;/m1./s1
SMIL Cl.CCOC(=O)[C@H]1C[C@H](C)CCN1C(=O)[C@@H](N)CCCNC(=N)N[N+](=O)[O-]
IUPAC navn ethyl (2R,4R)-1-[(2S)-2-amino-5-[(N-nitrocarbamimidoyl)amino]pentanoyl]-4-methylpiperidine-2-carboxylate;hydrochloride
Molekylvægt (g/mol) 408.88
Formel vægt 408.1888
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.