Få mere at vide
(2R,3S)-3-Amino-2-hydroxy-3-phenylpropanoic Acid Hydrochloride, Mikromol™
Beskrivelse
(2R,3S)-3-Amino-2-hydroxy-3-phenylpropanoic Acid Hydrochloride
Tekniske data
Tekniske data
| Kemisk navn eller materiale | (2R,3S)-3-Amino-2-hydroxy-3-phenylpropanoic Acid Hydrochloride |
| CAS | 132201-32-2 |
| Analyt- eller komponentnavne | (2R,3S)-3-Amino-2-hydroxy-3-phenylpropanoic Acid Hydrochloride |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C9 H11 N O3 . Cl H |
| Certificeringer/compliance | ISO 17025 |
| Synonym | (2R,3S)-3-Phenylisoserine hydrochloride, Benzenepropanoic acid, beta-amino-alpha-hydroxy-, hydrochloride (1:1), (alphaR,betaS)-, Benzenepropanoic acid, beta-amino-alpha-hydroxy-, hydrochloride, (alphaR,betaS)- (9CI), Benzenepropanoic acid, beta-amino-alpha-hydroxy-, hydrochloride, [R-(R*,S*)]- |
| InChI formel | InChI=1S/C9H11NO3.ClH/c10-7(8(11)9(12)13)6-4-2-1-3-5-6;/h1-5,7-8,11H,10H2,(H,12,13);1H/t7-,8+;/m0./s1 |
| SMIL | Cl.N[C@H]([C@@H](O)C(=O)O)c1ccccc1 |
| IUPAC navn | (2R,3S)-3-amino-2-hydroxy-3-phenylpropanoic acid;hydrochloride |
| Vis mere |