missing translation for 'onlineSavingsMsg'
Få mere at vide

(2R,3R)-3-Hydroxynorleucine, TRC

Artikelnummer. 30407531
Change view
Click to view available options
Mængde:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
30407531 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30407531 Supplier Toronto Research Chemicals Supplier No. TRCH949750100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale (2R,3R)-3-Hydroxynorleucine
CAS 59286-26-9
Molekylær formel C6H13NO3
Mængde 100 mg
Synonym (2R,3R)-2-Amino-3-hydroxy-hexanoic Acid, erythro-D-beta-Hydroxynorleucine
InChI formel InChI=1S/C6H13NO3/c1-2-3-4(8)5(7)6(9)10/h4-5,8H,2-3,7H2,1H3,(H,9,10)/t4-,5-/m1/s1
SMIL CCC[C@@H](O)[C@@H](N)C(=O)O
IUPAC navn (2R,3R)-2-amino-3-hydroxyhexanoic acid
Molekylvægt (g/mol) 147.17
Formel vægt 147.0895
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.