Få mere at vide
(2R)-3-Methyl-2-[pentanoyl[[2'-(1H-tetrazol-5-yl)biphenyl-4-yl]methyl]amino]butanoic Acid (Valsartan (R)-Enantiomer), Mikromol™
Beskrivelse
(2R)-3-Methyl-2-[pentanoyl[[2'-(1H-tetrazol-5-yl)biphenyl-4-yl]methyl]amino]butanoic Acid (Valsartan (R)-Enantiomer)
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Valsartan (R)-Enantiomer |
| CAS | 137862-87-4 |
| Analyt- eller komponentnavne | Valsartan (R)-Enantiomer |
| Forsendelsestilstand | Ice pack (-20°C) |
| Molekylær formel | C24 H29 N5 O3 |
| Certificeringer/compliance | ISO 9001 |
| Synonym | Valsartan Imp. A (EP), Valsartan USP Related Compound A, Valsartan USP RC A, (2R)-3-Methyl-2-[pentanoyl[[2'-(1H-tetrazol-5-yl)biphenyl-4-yl]methyl]amino]butanoic Acid, CGP 49309, D-Valsartan, N-(1-oxopentyl)-N-[[2'-(2H-tetrazol-5-yl)[1,1'-biphenyl]-4-yl]methyl]-D-Valine, Valsartan Related Compound A (USP), Valsartan Imp A (EP), Valsartan Related Compound A, Valsartan Impurity A |
| InChI formel | InChI=1S/C24H29N5O3/c1-4-5-10-21(30)29(22(16(2)3)24(31)32)15-17-11-13-18(14-12-17)19-8-6-7-9-20(19)23-25-27-28-26-23/h6-9,11-14,16,22H,4-5,10,15H2,1-3H3,(H,31,32)(H,25,26,27,28)/t22-/m1/s1 |
| SMIL | CCCCC(=O)N(Cc1ccc(cc1)c2ccccc2c3nnn[nH]3)[C@H](C(C)C)C(=O)O |
| IUPAC navn | (2R)-3-methyl-2-[pentanoyl-[[4-[2-(1H-tetrazol-5-yl)phenyl]phenyl]methyl]amino]butanoic acid |
| Vis mere |