Få mere at vide
(2R)-3,4-Dihydro-6-hydroxy-2,8-dimethyl-7-(methyl-d3)-2-[(4R,8R)-4,8,12-trimethyltridecyl]-2H-1-benzopyran-5-carboxylic-d3 Acid, TRC
Artikelnummer. 30376312
Skift visning
Mængde:
25 mg
Artikelnummer. Mængde
30376312 25 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30376312 Leverandør Toronto Research Chemicals Leverandørnr. TRCD45300725MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale (2R)-3,4-Dihydro-6-hydroxy-2,8-dimethyl-7-(methyl-d3)-2-[(4R,8R)-4,8,12-trimethyltridecyl]-2H-1-benzopyran-5-carboxylic-d3 Acid
CAS 842129-88-8
Molekylær formel C29H45D3O4
Mængde 25 mg
InChI formel InChI=1S/C29H48O4/c1-19(2)11-8-12-20(3)13-9-14-21(4)15-10-17-29(7)18-16-24-25(28(31)32)26(30)22(5)23(6)27(24)33-29/h19-21,30H,8-18H2,1-7H3,(H,31,32)/t20-,21-,29-/m1/s1/i5D3
SMIL [2H]C([2H])([2H])c1c(C)c2O[C@](C)(CCC[C@H](C)CCC[C@H](C)CCCC(C)C)CCc2c(C(=O)O)c1O
IUPAC navn (2R)-6-hydroxy-2,8-dimethyl-7-(trideuteriomethyl)-2-[(4R,8R)-4,8,12-trimethyltridecyl]-3,4-dihydrochromene-5-carboxylic acid
Molekylvægt (g/mol) 463.71
Formel vægt 463.3741