Få mere at vide
2H-Perfluoro-2-octenoic Acid, TRC
Artikelnummer. 30138471
Skift visning
Mængde:
1 mg
5 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30138471 10 mg
30012777 1 mg
30007627 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30138471 Leverandør Toronto Research Chemicals Leverandørnr. TRCD63017010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 70887-88-6
Molekylær formel C8 H2 F12 O2
Molekylvægt (g/mol) 358.08
Synonym 3,4,4,5,5,6,6,7,7,8,8,8-Dodecafluoro-2-octenoic acid, 2H-Perfluoro-2-octenoic acid, 3,4,4,5,5,6,6,7,7,8,8,8-Dodecafluorooct-2-enoic acid, FHUEA, 6:2 FTUCA
IUPAC navn (Z)-3,4,4,5,5,6,6,7,7,8,8,8-dodecafluorooct-2-enoic acid
SMIL OC(=O)\C=C(/F)\C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F
Tekniske data
Kemisk navn eller materiale 2H-Perfluoro-2-octenoic acid
Mængde 10 mg
InChI formel InChI=1S/C8H2F12O2/c9-2(1-3(21)22)4(10,11)5(12,13)6(14,15)7(16,17)8(18,19)20/h1H,(H,21,22)/b2-1-
Formel vægt 357.9863