missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2H-Chromene-3-carboxylic acid, 90+%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
10 g
Specifications
Assay Percent Range | 90% |
Quantity | 250 mg |
Formula Weight | 176.17 |
Percent Purity | 90% |
Chemical Name or Material | 2H-Chromene-3-carboxylic acid |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'