missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
2H,2H,3H,3H-Perfluorooctanoic Acid, TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | 2H,2H,3H,3H-Perfluorooctanoic Acid |
| CAS | 914637-49-3 |
| Renhedsgrad noter | GC |
| Molekylær formel | C8 H5 F11 O2 |
| Mængde | 250 mg |
| Synonym | 2H,2H,3H,3H-Perfluorooctanoic Acid, Octanoic acid','4,4,5,5,6,6,7,7,8,8,8-undecafluoro-, 4,4,5,5,6,6,7,7,8,8,8-Undecafluorooctanoic acid, 2H,2H,3H,3H-Perfluorooctanoic acid, 5:3 Fluorotelomer carboxylic acid, 5:3 FTCA, 5:3 FTCA, 4,4,5,5,6,6,7,7,8,8,8-Undecafluoro-octanoic Acid, 4,4,5,5,6,6,7,7,8,8,8-Undecafluorooctanoic Acid |
| InChI formel | InChI=1S/C8H5F11O2/c9-4(10,2-1-3(20)21)5(11,12)6(13,14)7(15,16)8(17,18)19/h1-2H2,(H,20,21) |
| SMIL | OC(=O)CCC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F |
| IUPAC navn | 4,4,5,5,6,6,7,7,8,8,8-undecafluorooctanoic acid |
| Molekylvægt (g/mol) | 342.11 |
| Vis mere |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?