Få mere at vide
2H-2-Ethyl-d5 Candesartan, TRC
Artikelnummer. 30370882
Skift visning
Mængde:
2.5 mg
Artikelnummer. Mængde
30370882 2.5 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30370882 Leverandør Toronto Research Chemicals Leverandørnr. TRCE9008672.5MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2H-2-Ethyl-d5 Candesartan
CAS 1246820-58-5
Molekylær formel C26H19D5N6O3
Mængde 2.5 mg
Synonym 2-Ethoxy-1-[[2'-[2-(ethyl-d5)-2H-tetrazol-5-yl][1,1'-biphenyl]-4-yl]methyl]-1H-benzimidazole-7-carboxylic Acid
InChI formel InChI=1S/C26H24N6O3/c1-3-32-29-24(28-30-32)20-9-6-5-8-19(20)18-14-12-17(13-15-18)16-31-23-21(25(33)34)10-7-11-22(23)27-26(31)35-4-2/h5-15H,3-4,16H2,1-2H3,(H,33,34)/i1D3,3D2
SMIL [2H]C([2H])(C([2H])([2H])[2H])N(N=N1)N=C1C2=CC=CC=C2C(C=C3)=CC=C3CN(C(OCC)=N4)C5=C4C=CC=C5C(O)=O
IUPAC navn 2-ethoxy-1-((2'-(2-(ethyl-d5)-2H-tetrazol-5-yl)-[1,1'-biphenyl]-4-yl)methyl)-1H-benzo[d]imidazole-7-carboxylic acid
Molekylvægt (g/mol) 473.54
Formel vægt 473.22