Få mere at vide
(2E)-Octadecenal (>90%), TRC
Artikelnummer. 30137607
Skift visning
Mængde:
50 mg
250 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30137607 250 mg
30138587 500 mg
30012746 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30137607 Leverandør Toronto Research Chemicals Leverandørnr. TRCO233700250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 51534-37-3
Molekylær formel C18 H34 O
Molekylvægt (g/mol) 266.46
Synonym Koiganal I, trans-2-Octadecenal, (E)-2-Octadecenal;
IUPAC navn (E)-octadec-2-enal
SMIL CCCCCCCCCCCCCCC\C=C\C=O
Tekniske data
Kemisk navn eller materiale (2E)-Octadecenal
Mængde 250 mg
InChI formel InChI=1S/C18H34O/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19/h16-18H,2-15H2,1H3/b17-16+
Formel vægt 266.261